missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Acrylic Vacuum Desiccators

$2,101.55 - $2,299.55

Specifications

Width (English) Opening 9.72 in.
Width (Metric) Opening 24.7cm
Material Acrylic
Width (Metric) 21cm
Product Type Vacuum Desiccator
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
03-410-570 Bel-Art™
F420800000
Removable cover, silicone gasket Each for $2,101.55
03-410-571 Bel-Art™
F420800115
Removable cover, silicone gasket and plug socket Each for $2,299.55
Specifications Description & Specifications

Specifications

9.72 in.
Acrylic
Vacuum Desiccator
21cm
24.7cm
24.7cm
21cm
21cm
9.72 in.