Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ 50mL Conical Tube Holders


Leave both hands free for manipulation of 50mL sample tubes

Manufacturer: bel-art™  F187950000

 View more versions of this product

Catalog No. 08-647-121

  • / Pack of 5 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Conical tube
Material Polystyrene
Autoclavable No
Color White
For Use With (Equipment) Conical tube
Product Type Tube Holder, Non-Grip Style
Type Tube Holder, Non-Grip Style