Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ 50mL Conical Tube Holders


Leave both hands free for manipulation of 50mL sample tubes

Manufacturer: bel-art™  F187950001

 View more versions of this product

Catalog No. 08-647-120

  • / Pack of 5 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Conical tube
Product Type Tube Holder, Grip Style
Material Polystyrene
Autoclavable No
Type Tube Holder, Grip Style
For Use With (Equipment) Conical tube
Color Blue