missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ 50mL Centrifuge Tube Refrigerator Rack

Provides efficient and extended cooling of twenty-eight 50mL centrifuge tubes in a chilled bath

Manufacturer:  Bel-Art™ 189050051

 View more versions of this product

Catalog No. 03-410-625


 

Specifications Description & Specifications

Specifications

Polypropylene
15cm
No
5.875 in.
Cover
Centrifuge tube ice tray
Promotions

Promotions