Please login to your online account to display your discounted pricing
Bel-Art H-B™ DURAC™ Single Channel Timer with Ten Button Direct Input
Ten button timer allows direct input for faster programming of desired time
$41.32 - $41.32
Specifications
Battery Type Two 1.5V, type AAA or equivalent (included)
Product Type Single Channel Timer
Resolution One second
Timer ±Five seconds/day
View More Specs
Includes:  Two AAA batteries
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
13-202-732
fisher scientific
B617003500
Provided with Certificate of Calibration. This certificate indicates calibration using NIST procedures and is in compliance with ANSI/NCSL Z540-1. 10/Cs. Each for $41.32
Description & Specifications
Specifications
Battery Type Two 1.5V, type AAA or equivalent (included)
Product Type Single Channel Timer
Resolution One second
Timer ±Five seconds/day
  • Timing capacity: 99 minutes: 99 seconds
  • Count direction: up/down
  • Last time set memory
  • Audible and visual alarms; displays elapsed time after countdown
  • Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment
  • Includes multi-language instructions (English, French, German, Italian, Portuguese and Spanish)