Please login to your online account to display your discounted pricing
Bel-Art H-B™ DURAC™ Four Channel Timer with White Board
Four channel timer has built in white board that helps keep track of time and other critical information
$85.89 - $85.89
Specifications
Battery Type Two 1.5V, type AAA or equivalent (included)
Product Type Four Channel Timer with White Board
Resolution One second
Timer ±Five seconds/day
View More Specs
Includes:  Two AAA batteries, multi-language instructions (English, French, German, Italian, Portuguese and Spanish)
Products
Catalog Number Mfr. No. Includes Quantity Price Quantity    
13-202-731
fisher scientific
B617003700
Timer with White Board 10/Cs. Each for $85.89
Description & Specifications
Specifications
Battery Type Two 1.5V, type AAA or equivalent (included)
Product Type Four Channel Timer with White Board
Resolution One second
Timer ±Five seconds/day
  • Audible alarm for each channel
  • Built in white board and dry erase marker with eraser tip (included) helps keep track of crucial information
  • Programs: hours/minutes; Displays: hours/minutes/seconds
  • Switchable timing capacity: 23 hours: 59 minutes / 59 minutes
  • Count direction: up/down; displays elapsed time after countdown
  • Last time set memory
  • Key press confirmation feature gives an audible indication of data entry
  • Lay flat on a bench, use the flip-open stand for a vertical view, or use magnetic back for metal surface attachment