missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Behenic Anhydride 97.0+%, TCI America™
$173.37 - $538.71
Chemical Identifiers
| CAS | 55726-23-3 |
|---|---|
| Molecular Formula | C44H86O3 |
| Molecular Weight (g/mol) | 663.17 |
| MDL Number | MFCD00010452 |
| InChI Key | IJVLPVWXJLKHID-UHFFFAOYSA-N |
| Synonym | Docosanoic Anhydride |
| PubChem CID | 566696 |
| IUPAC Name | docosanoyl docosanoate |
| SMILES | CCCCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCCCC |
Chemical Identifiers
| 55726-23-3 | |
| 663.17 | |
| IJVLPVWXJLKHID-UHFFFAOYSA-N | |
| 566696 | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCCCC |
| C44H86O3 | |
| MFCD00010452 | |
| Docosanoic Anhydride | |
| docosanoyl docosanoate |
Specifications
| 55726-23-3 | |
| White-Yellow | |
| MFCD00010452 | |
| Docosanoic Anhydride | |
| CCCCCCCCCCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCCCCCCCCCC | |
| 663.17 | |
| 663.17 | |
| Crystalline Powder |
| 84°C | |
| C44H86O3 | |
| 1 g | |
| IJVLPVWXJLKHID-UHFFFAOYSA-N | |
| docosanoyl docosanoate | |
| 566696 | |
| ≥97.0% (GC) | |
| Behenic Anhydride |
Safety and Handling
TSCA : No