Please login to your online account to display your discounted pricing

Begacestat, Tocris™

γ-secretase inhibitor; lowers Aβ40 and Aβ42 levels

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material Begacestat
Assay Percent Range >98%
Molecular Formula C9H8ClF6NO3S2
Formula Weight 391.74
Synonym 5-Chloro-N-[(1S)-3, 3, 3-trifluoro-1-(hydroxymethyl)-2-(trifluoromethyl)propyl]-2-thiophenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

428350

 
r & d systems
RDS428350
50mg Each for $997.23

428310

 
r & d systems
RDS428310
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 391.74
Quantity 50mg
Chemical Name or Material Begacestat
Assay Percent Range >98%
Molecular Formula C9H8ClF6NO3S2
CAS 769169-27-9
Synonym 5-Chloro-N-[(1S)-3, 3, 3-trifluoro-1-(hydroxymethyl)-2-(trifluoromethyl)propyl]-2-thiophenesulfonamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C