Please login to your online account to display your discounted pricing

Beclin-1 Activator I, TAT-Beclin-1, Calbiochem™, EMD Millipore

A cell-permeable Atg6/Beclin1-derived autophagy-inducing peptide, Peptide Sequence: H-Tyr-Gly-Arg-Lys-Lys-Arg-Arg-Gln-Arg-Arg-Arg-Gly-Gly-Thr-Asn-Val-Phe-Asn-Ala-Thr-Phe-Glu-Ile-Trp-His-Asp-Gly-Glu-Phe-Gly-Thr-OH; Packaged under inert gas

$362.90 - $362.90

Chemical Identifiers

Assay Percent Range ≥97.0%
Chemical Name or Material Beclin-1 Activator I, TAT-Beclin-1
Molecular Formula C164H251N57O45
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5060480001

 
emd millipore corporation
5060480001
Each for $362.90
Description & Specifications

Specifications

Quantity 10mg
Molecular Formula C164H251N57O45
Synonym H2N-YGRKKRRQRRR-GG-TNVFNATFEIWHDGEFGT-CO2H; Atg6 Activator I; Beclin1-GAPR-1 Interaction Blocker I; Vps30 Activator I; Autophagy Inducer IV
Chemical Name or Material Beclin-1 Activator I, TAT-Beclin-1
Solubility Information Soluble in DMSO (50mg/mL) or H2O (50mg/mL)
Recommended Storage 2°C to 8°C (Protect from light).
Purity Grade Notes by HPLC
Assay Percent Range ≥97.0%
Safety and Handling

2°C to 8°C (Protect from light).