Please login to your online account to display your discounted pricing

Beckman Coulter™ StatSpin OvaTube Centrifugation Tubes DFS Item

$929.77 - $929.77

Specifications

For Use With (Application) Parasite testing
Sample Type Parasite
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

22-269-248

DFS Item
 
beckman coulter
SSOT250
250/Box Case of 250 Each for $929.77
Description & Specifications

Specifications

For Use With (Application) Parasite testing
Sample Type Parasite