Please login to your online account to display your discounted pricing

Beckman Coulter™ Gastroccult™ Gastric Sample Occult Blood Test Kit

Two tests in one

Manufacturer: beckman coulter  66115A

 View all options for this product

Catalog No. SK-66115


Description & Specifications


Clia Complexity Waived
CPT Code 87723
For Use With (Equipment) Gastric occult Blood and pH test kit
Quantity 6 x 15mL
Product Type Gastroccult developer

  • CLIA Complexity: Waived
  • CPT Codes: 87723 for occult blood, 83986 for pH
  • Slide test for detecting occult blood and determining pH in gastric samples, both aspirate and vomitus, on the same test card
  • On-slide Performance Monitor assures that the occult blood test is performing to product specifications
  • Scale of pH test ranges from a clinically useful 1.3 to 7.8
  • Both test results are available within one minute
  • Test cards, developer, and applicator straws sold separately