missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BDE No 85 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™
Description
Environmental
Specifications
Specifications
| CAS | 182346-21-0 |
| Flash Point | -12°C - Closed Cup |
| Linear Formula | C12H5 Br5 O |
| MDL Number | MFCD08460513 |
| Synonym | 2,2 ′,3,4 ′,4-Pentabromodiphenyl ether solution; 2,2 ′,3,4 ′,4-PentaBDE; PBDE 85 |
| Type | Analytical Standard |
| Quantity | 1 mL |
| Formula Weight | 564.69 |
| Grade | Analytical Standard |
| Concentration or Composition (by Analyte or Components) | 50 μg/mL in isooctane |