Please login to your online account to display your discounted pricing

BDCS, silylation reagent, AcroSeal™, ACROS Organics™


Quantity 5mL
Packaging AcroSeal glass bottle

Manufacturer: thermo fisher scientific  200940050

 View more versions of this product

Catalog No. AC200940050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Solution
Formula Weight 218.8
Quantity 5mL
Color Clear light yellow
Fieser 07,184; 01,489; 03,157; 08,255
Chemical Name or Material BDCS
Infrared Spectrum Authentic
Packaging AcroSeal glass bottle
Refractive Index 1.4400 to 1.4420 (20°C, 589nm)
Molecular Formula C6H15ClSi·C3H4N2
MDL Number MFCD00000501
CAS 1185092-02-7
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Contact with water liberates toxic gas.

Specifications

Chemical Name or Material BDCS
CAS 1185092-02-7
Molecular Formula C6H15ClSi·C3H4N2
Formula Weight 218.8
MDL Number MFCD00000501
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Contact with water liberates toxic gas.

SDS & Safety Documents