missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BDCS, silylation reagent, AcroSeal™, ACROS Organics™
Chemical Identifiers
CAS | 1185092-02-7 |
---|---|
Molecular Formula | C6H15ClSi·C3H4N2 |
Molecular Weight (g/mol) | 218.8 |
MDL Number | MFCD00000501 |
InChI Key | IFLPAOFWVBWJPG-UHFFFAOYSA-N |
Synonym | bdcs silylation reagent, bdcs, silylation reagent, acroseal, him; t-butyldimethylchlorosilane, imidazole; t-butyldimethylchlorosilane, t-butyl-dimethylsilyl chloride-imidazole, tert-butyl-chloro-dimethylsilane; 1h-imidazole, tert-butyldimethylchlorosilane imidazole |
PubChem CID | 2724131 |
IUPAC Name | tert-butyl-chloro-dimethylsilane;1H-imidazole |
SMILES | CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC200941000
|
Thermo Fisher Scientific
200941000 |
100mL | AcroSeal™ Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC200948000
|
Thermo Fisher Scientific
200948000 |
800mL | AcroSeal™ Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
1185092-02-7 | |
218.8 | |
IFLPAOFWVBWJPG-UHFFFAOYSA-N | |
2724131 | |
CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1 |
C6H15ClSi·C3H4N2 | |
MFCD00000501 | |
bdcs silylation reagent, bdcs, silylation reagent, acroseal, him; t-butyldimethylchlorosilane, imidazole; t-butyldimethylchlorosilane, t-butyl-dimethylsilyl chloride-imidazole, tert-butyl-chloro-dimethylsilane; 1h-imidazole, tert-butyldimethylchlorosilane imidazole | |
tert-butyl-chloro-dimethylsilane;1H-imidazole |
Specifications
0.9590g/mL | |
Authentic | |
1.4400 to 1.4420 | |
Yellow | |
1185092-02-7 | |
MFCD00000501 | |
bdcs silylation reagent, bdcs, silylation reagent, acroseal, him; t-butyldimethylchlorosilane, imidazole; t-butyldimethylchlorosilane, t-butyl-dimethylsilyl chloride-imidazole, tert-butyl-chloro-dimethylsilane; 1h-imidazole, tert-butyldimethylchlorosilane imidazole | |
CC(C)(C)[Si](C)(C)Cl.C1=CN=CN1 | |
218.8 | |
218.8 | |
BDCS |
58°C | |
AcroSeal™ Glass bottle | |
0.959 | |
100mL | |
C6H15ClSi·C3H4N2 | |
07,184; 01,489; 03,157; 08,255 | |
IFLPAOFWVBWJPG-UHFFFAOYSA-N | |
tert-butyl-chloro-dimethylsilane;1H-imidazole | |
2724131 | |
Solution |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of damaging the unborn child.
Contact with water liberates toxic gas.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Danger