BD Veritor™ System for the Rapid Detection of Flu A+B
Unique adaptive-read technology helps reduce false-positive results
$139.13 - $2,165.03
Specifications
For Use With (Application) Detection of Flu A + B
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Quantity Price Quantity  
Catalog Number Mfr. No. Description Quantity Price Quantity  
B256055 BD
B256055
System Reader Each Each for $1,110.18
22-351-287 BD
B256051
Control Swab Set, 10 pr. Each Pack of 10 Each for $139.13
B256041 BD
B256041
Flu A +  B Test Kit Each Pack of 30 Each for $2,165.03
Specifications
Description
  • Requires minimal hands-on time
  • Easy sample processing – Unitized tube containing correct volume of process reagent facilitates workflow
  • Ready in minutes – Test device is ready to insert into reader 10 minutes after sample is added
  • Insert and read – Simple one-touch button readies reader for test device insertion
  • Results delivered – Once test device is inserted in reader, an objective, digital test result is displayed in 10 seconds
  • Cleared for use with nasal swabs and nasopharyngeal (NP) swabs
  • CLIA Complexity: Waived
Description & Specifications
Specifications
Detection of Flu A + B