Please login to your online account to display your discounted pricing

BD Veritor System RSV (Respiratory Syncytial Virus) Test Kit


Provides objective results on an easy to read digital display, unlike current rapid RSV tests, which requires subjective visual test interpretation to determine positive or negative test result

Manufacturer: bd  B256042

 View more versions of this product

Catalog No. B256042

  • / Pack of 30 Each
 

Description & Specifications

Specifications

Sample Type Nasal Aspirate
Product Type RSV Test Kit
Quantity 30 Tests
Clia Complexity Moderate
Detectable Analytes RSV
Test Time 10 sec.

  • Intended for in vitro diagnostic use to aid in diagnosis of RSV infections in infants and pediatric patients under age of 20 years
  • For use with BD Veritor System Reader