Please login to your online account to display your discounted pricing

BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Hemogard Closure


Use for serum determinations in chemistry, serology and immunohematology

Manufacturer: bd  B367812

 View more versions of this product

Catalog No. 02-685-111

  • / Pack of 100 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Additive Clot activator, Silicone coated
Capacity (Metric) 4mL
Diameter (Metric) Outer 13 x 75mm
Color Red
Material Plastic
Closure Type Hemogard