Please login to your online account to display your discounted pricing

BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Plastic Serum Tubes, Silicone-Coated, with Conventional Stopper


Use for serum determinations in chemistry, serology and immunohematology

Manufacturer: bd  B367820

 View more versions of this product

Catalog No. 02-683-94B

  • / Pack of 100 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Material Plastic
Additive Clot activator, Silicone coated
Color Red
Diameter (Metric) Outer 16 × 100mm
Capacity (Metric) 10.0 mL
Closure Type Conventional