BD Vacutainer™ Venous Blood Collection Tubes: Vacutainer Plus™ Glass Serum Tubes, Silicone-Coated, with Conventional Stopper
Use for serum determinations in chemistry, serology and immunohematology
$41.58 - $41.58
Specifications
Additive Silicone coated
Color Red
Closure Type Conventional
Material Glass
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
02-685A BD
B366430
16 x 100mm; 10.0mL Pack of 100 Each for $41.58
Case of 10 Pack for $415.80
Specifications
Description
  • Sterile vacuum blood collection tubes for serum determination
  • Spray-coated silica interior
  • With conventional tube stopper
Evacuated Tubes Simplify Blood Collection
  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded
Description & Specifications
Specifications
Silicone coated
Red
Conventional
Glass