Please login to your online account to display your discounted pricing

BD Vacutainer™ Venous Blood Collection Tubes: SST™ Serum Separation Tubes: Conventional Stopper


Ideal for use in serum determinations in chemistry

Manufacturer: bd  B367987

 View more versions of this product

Catalog No. 02-683-95

  • / Pack of 100 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Applications For use in serum determinations in chemistry
Material Plastic
Additive Polymer gel/Silica activator
Color Red/Gray
Capacity (Metric) 16 x 100 mm, 7.5 mL
Closure Type Conventional