BD Vacutainer™ Urine Collection Kits: Urinalysis
Reduce the need to retest, relabel, and/or recollect specimens
Manufacturer:  BD 364943
 View more versions of this product
Catalog No. 14-827-91
Description
Description
  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
Urinalysis Products
  • Include BD™ proprietary mercury-free Urinalysis Preservative Tube, which provides compliance with both the Memorandum of Understanding for the U.S. EPA and the AHA
  • Help to maintain urine sample integrity for up to 72 hours at room temperature
  • Minimum and maximum fill lines (7-8mL) ensure proper additive-to-urine ratio
Specifications
Specifications
Urine Collection Kit
Healthcare
Urinalysis preservative, transfer device, evacuated conical bottom tube
For UA chemistry Urine testing
Room temperature
Sterile
0.27 oz.
8mL
Plastic
Yes
0.27 oz., 8mL
Documents