missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Urine Collection Kits: Urinalysis

Reduce the need to retest, relabel, and/or recollect specimens

Manufacturer:  BD 364990

 View more versions of this product

Catalog No. 02-683-127




Description

Description

  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes

Urinalysis Products

  • Include BD™ proprietary mercury-free Urinalysis Preservative Tube, which provides compliance with both the Memorandum of Understanding for the U.S. EPA and the AHA
  • Help to maintain urine sample integrity for up to 72 hours at room temperature
  • Minimum and maximum fill lines (7-8mL) ensure proper additive-to-urine ratio

Specifications

Specifications

0.33 oz.
Transfer device, evacuated round bottom tube
For UA chemistry Urine testing
Room temperature
Sterile
10mL
Plastic
Yes
0.33 oz., 10mL
SDS
Documents