Please login to your online account to display your discounted pricing

BD Vacutainer™ Plastic Blood Collection Tubes with Sodium Citrate


Evacuated glass blood collection tubes for routine coagulation studies

Manufacturer: bd  B363083

 View more versions of this product

Catalog No. 02-683-172

  • / Pack of 100 Each
  • / Case of 10 Pack
 

Description & Specifications

Specifications

Product Type BD Vacutainer™ Plus Draw Tube
Additive Buffered Sodium Citrate, 3.2%
Color Blue
Diameter (Metric) Outer 13 x 75mm
Material Plastic
Capacity (Metric) 2.7mL
Closure Type Hemogard