missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Vacutainer™ Glass Blood Collection Tubes: Buffered Sodium Citrate

Evacuated glass blood collection tubes for routine coagulation studies

$35.61 - $35.61

Specifications

Applications Blood Coagulation
Color Blue
Diameter (Metric) Outer 13mm
Capacity (Metric) 4.5mL
Closure Type Hemogard
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Additive Price Quantity  
Catalog Number Mfr. No. Additive Price Quantity  
02-688-26 BD
B369714
Sodium citrate (0.105M, 3.2%) Pack of 100 Each for $35.61
Case of 10 Pack for $356.10
Specifications

Description

  • BD Vacutainer Citrate™ tubes with 3.2% buffered sodium citrate solution are used for routine coagulation studies

  • Blood is drawn directly from the vein into the evacuated sterile collection tubes
  • Only one venipuncture is needed to draw multiple samples
  • Tubes can be changed while needle and plastic holder remain in place on patient's arm
  • Closures are color coded

Hemogard Closures

  • Safety shield designed for safer tube handling
  • Rubber stopper is recessed inside the plastic shield, so any drops left by a blood collection needle remain isolated from potential contact
  • Shield is designed to protect lab personnel from direct contact with blood on the stopper or around the outer rim of the tube, as well as from blood splattering upon opening the tube

Description & Specifications

Specifications

Blood Coagulation
13mm
Hemogard
75mm
Blue
4.5mL
Paper