BD Sedi-Stain™ Concentrated Stain for Urinary Sediment
No mixing of solutions is required
$42.60 - $99.45
Specifications
For Use With (Application) Highly selective formula stains Blood cells, casts and other formed elements in Urine sediment in a distinctive fashion that permits rapid and accurate identification
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
02-659 BD
B421571
60mL Pack for $99.45
Case of 10 Pack for $994.50
23-590992 BD
B421570
12.5mL Pack for $42.60
Case of 10 Pack for $426.00
Specifications
Description
  • Stabilized modification of the Sternheimer-Mablin urinary stain
  • Highly selective formula stains blood cells, casts, and other formed elements in urine sediment in a distinctive fashion that permits rapid and accurate identification
  • Sedi-Stain is used directly from the dropper bottle and will remain stable indefinitely
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.
Description & Specifications
Specifications
Highly selective formula stains Blood cells, casts and other formed elements in Urine sediment in a distinctive fashion that permits rapid and accurate identification