BD SafetyGlide™ Needles
Needles feature patented Activation-Assist™ Technology for fast and easy, single-handed needle-tip shielding immediately after injection.
Manufacturer:  BD 305900
 View more versions of this product
Catalog No. 14-829-13
Description
Description
  • This technology provides versatility and minimal change in technique
Specifications
Specifications
Black
Yes
General and hypodermic use
38.1mm
Sterile
22 G
Shielding mechanism
1.5 in.
Regular Bevel
Shielding Hyperdermic Needle
Documents