Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) w Lecithin, Polysorbate 80 (LP80), Penicillinase

$852.08 - $896.56

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Format Liquid
For Use With (Application) Rodac™ snap lid has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate
Manufacturing Origin United States
View More Specs
Products
Catalog Number Mfr. No. Shelf Life Media Type Packaging Quantity Price Quantity    

B215273

 
bd
B215273
112 Days Trypticase™ Soy Agar with Lecithin and Polysorbate 80 and Penicillinase VHP Resistant Triple Wrapping Isolator Pack 100/Pk. Pack of 100 Each for $891.54

B215274

 
bd
B215274
98 Days Trypticase™ Soy Agar with Lecithin, Polysorbate 80 and Penicillinase Sterile Double Wrapping Plate 100/Pk. Pack of 100 Each for $896.56

B215279

 
bd
B215279
98 Days Trypticase™ Soy Agar with Lecithin, Polysorbate 80 and Penicillinase Sterile Double Wrapping Plate 10/Pk. Pack of 10 Each for $852.08
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Format Liquid
For Use With (Application) Rodac™ snap lid has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate
Manufacturing Origin United States

The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.

The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:

  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage

Safety and Handling

112 Days