Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) w Lecithin, Polysorbate 80 (LP80), Penicillinase


Manufacturer: bd  B215279

 View more versions of this product

Catalog No. B215279

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Sterile Yes
Format Liquid
Media Type Trypticase™ Soy Agar with Lecithin, Polysorbate 80 and Penicillinase
Color Light Beige
Packaging Sterile Double Wrapping Plate
Certifications/Compliance ISO/DIS, 11014
Manufacturing Origin United States
Shelf Life 98 Days
For Use With (Application) Rodac™ snap lid has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate
Accessories

98 Days