Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)


Manufacturer: bd  B215284

 View more versions of this product

Catalog No. B215284

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Sterile Yes
Format Liquid
Certifications/Compliance ISO/DIS, 11014
Shelf Life 112 Days
Color Light Beige
Packaging VHP Resistant Triple Wrapping Isolator Pack
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbate 80
Manufacturing Origin United States
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Accessories

112 Days

SDS & Safety Documents