Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) with Lecithin and Polysorbate 80 (LP80)


Manufacturer: bd  B215277

 View more versions of this product

Catalog No. B215277

  • / Pack of 20 Each
 

Description & Specifications

Specifications

Quantity 20/Pk.
Format Liquid
Packaging Single Wrapping, Aseptic Fill
Shelf Life 112 Days
Certifications/Compliance ISO, 9000
Color Light Beige
For Use With (Application) Cultivation of fastidious microorganisms and visualization of hemolytic reactions
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin and Polysorbate 80