Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID Sabouraud Dextrose Agar (SDA) with Lecithin and Polysorbate 80 (LP80)


Manufacturer: bd  B215283

 View more versions of this product

Catalog No. B215283

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Sterile Yes
Packaging Sterile Double Wrapping Plate
Media Type Sabouraud Dextrose Agar with Lecithin and Polysorbate 80
Shelf Life 112 Days
Color Light Beige
Includes Rodac™ snap lid
Certifications/Compliance ISO/DIS, 11014
Format Liquid
Manufacturing Origin United States
Accessories

112 Days