Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID D/E Neutralizing Agar


Manufacturer: bd  B215271

 View more versions of this product

Catalog No. B215271

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Sterile Yes
Packaging VHP Resistant Triple Wrapping Isolator Pack
Certifications/Compliance ISO/DIS, 11014
Weight (Metric) 770g
Shelf Life 112 Days
Color Purple
Manufacturing Origin United States
Format Liquid
Includes Rodac™ snap lid
Media Type D/E neutralizing Agar
Accessories

112 Days