Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID D/E Neutralizing Agar

$366.49 - $1,216.10

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Purple
Format Liquid
Media Type D/E neutralizing Agar
Includes Rodac™ snap lid
View More Specs
Products
Catalog Number Mfr. No. Weight (Metric) Packaging Quantity Price Quantity    

B215271

 
bd
B215271
770g VHP Resistant Triple Wrapping Isolator Pack 10/Pk. Pack of 10 Each for $366.49

B215280

 
bd
B215280
2720g Sterile Double Wrapping Plate 100/Pk. Pack of 100 Each for $977.64

B215281

 
bd
B215281
900g Sterile Double Wrapping Plate 10/Pk. Pack of 10 Each for $1,216.10
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Purple
Format Liquid
Media Type D/E neutralizing Agar
Includes Rodac™ snap lid
Manufacturing Origin United States
Shelf Life 112 Days

The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.

The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:

  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage

Safety and Handling

112 Days