Please login to your online account to display your discounted pricing

BD RODAC™ SNAP LID D/E Neutralizing Agar


Manufacturer: bd  B215281

 View more versions of this product

Catalog No. B215281

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Quantity 10/Pk.
Sterile Yes
Weight (Metric) 900g
Format Liquid
Color Purple
Shelf Life 112 Days
Media Type D/E neutralizing Agar
Packaging Sterile Double Wrapping Plate
Certifications/Compliance ISO/DIS, 11014
Includes Rodac™ snap lid
Manufacturing Origin United States
Accessories

112 Days