missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Probetec™ ET CT/GC Specimen Collection and Dry Transport System

Offers a convenient way to collect and transport specimens from the patient to the laboratory

$521.54 - $521.54

Specifications

For Use With (Equipment) BD Probetec™ ET CT or GC amplified DNA assays
Quantity 100 Pack
Sterile Yes
Storage Requirements 15 to 30°C
View More Specs

Includes: Male Urethral Kit: Each unit includes one rayon-tipped swab on a flexible and extendable wire in a tube
Endocervical Kit: Each unit contains one polyurethane-tipped swab in a tube and a large rayon-tipped cleaning swab


Products ${productFamilyLength}
Catalog Number Mfr. No. Closure Color Includes Product Type Price Quantity  
Catalog Number Mfr. No. Closure Color Includes Product Type Price Quantity  
22-370-166 BD
B440461
Blue One Rayon-tipped swab on a flexible and extendable wire in a tube Male Urethral Specimen Collection and Dry Transport Kit Pack of 100 Each for $521.54
Specifications

Description

BD ProbeTec™ ET CT/GC Amplified DNA Assay Specimen Collection and DRY TRANSPORT Kit

  • Sterile, self-contained, disposable systems
  • Specialized swabs for collection of urethral or endocervical specimens
  • Swabs facilitate obtaining specimens and maintaining the organisms for nucleic acid amplification
  • For use with the BD ProbeTec ET Chlamydia trachomatis and Neisseria gonorrhoeae Amplified DNA assays on the BD ProbeTec ET System

Endocervical Kit

  • Designed for the cleaning and collection of endocervical specimens
  • Large cleaning swab is included to clean the cervical os of blood and mucus prior to collection
  • Polyurethane-tipped swab on a plastic shaft
Male Urethral Kit
  • Designed for the collection of male urethral specimens
  • Flexible aluminum wire and small rayon-tipped swab facilitate obtaining the specimen
  • Plastic handle housing the aluminum wire and secured in the cap provides a slide action for extension of the swab

Description & Specifications

Specifications

BD Probetec™ ET CT or GC amplified DNA assays
100 Pack
Yes
15 to 30°C