BD PrecisionGlide™ Single-use Needles: Short Bevel - Thin Wall
Manufacturer:  BD 305188
 View more versions of this product
Catalog No. 14-821-14G
Description
Description
  • PrecisionGlide stainless-steel needles with translucent, color-coded, polypropylene hubs
  • Can also be sterilized with ethylene oxide (EtO) in their individual, easy-to-peel blister package
Order Info
Needles are autoclavable after removal from package.
Specifications
Specifications
Brown
19 G
1 in.
25mm
Sterile
Yes
Facilitate the injection of high-viscosity fluids
Stainless steel
Bevel
Thin Wall Syringe Needle
Documents