missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD PrecisionGlide™ Single-use Needles: Short Bevel - Thin Wall

$9.84 - $98.44

Specifications

Autoclavable Yes
Color Brown
For Use With (Application) Facilitate the injection of high-viscosity fluids
Gauge 19 ga
Material Stainless Steel
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Length (Metric) Needle Price Quantity  
Catalog Number Mfr. No. Length (Metric) Needle Price Quantity  
14-821-14H BD
305189
38mm Pack of 100 Each for $9.84
Case of 10 Pack for $98.40
14-821-14G BD
305188
25mm Case of 1000 Each for $98.44
Specifications

Description

  • PrecisionGlide stainless-steel needles with translucent, color-coded, polypropylene hubs
  • Can also be sterilized with ethylene oxide (EtO) in their individual, easy-to-peel blister package

Order Info

Needles are autoclavable after removal from package.

Description & Specifications

Specifications

Yes
Facilitate the injection of high-viscosity fluids
Stainless Steel
Yes
Brown
19 ga
Bevel
Thin Wall Syringe Needle