BD PrecisionGlide™ Single-use Needles: Regular Bevel - Regular Wall
Manufacturer:  BD B305128
 View more versions of this product
Catalog No. 14-821-13A
Description
Description
  • PrecisionGlide stainless-steel needles with translucent, color-coded, polypropylene hubs
  • Can also be sterilized with ethylene oxide (EtO) in their individual, easy-to-peel blister package
  • Regular wall, regular bevel
Order Info
Needles are autoclavable after removal from package.
Specifications
Specifications
Beige
30 G
1 in.
25mm
Sterile
Yes
Facilitate the injection of high-viscosity fluids
Stainless steel
Bevel
Syringe Needle
Documents
Accessories