BD PrecisionGlide™ Single-use Needles: Fill Needles - Regular Bevel - Thin Wall
Manufacturer: BD 305185
Description
- PrecisionGlide stainless-steel needles with translucent, color-coded, polypropylene hubs
- Can also be sterilized with ethylene oxide (EtO) in their individual, easy-to-peel blister package
Order Info
Needles are autoclavable after removal from package.
Specifications
Stainless Steel | |
Thin Wall Syringe Needle | |
Pink | |
Facilitate the injection of high-viscosity fluids |
Sterile | |
Autoclavable | |
18 G | |
1.5 in. |