Please login to your online account to display your discounted pricing

BD Difco™ Peptone Water

For cultivation of nonfastidious organisms and studying carbohydrate fermentation

$461.38 - $461.38

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Liquid
For Use With (Application) Sterility testing, non-sterile product testing, food, dairy and Water testing
Packaging Bottle
Shelf Life 365 Days
View More Specs
Products
Catalog Number Mfr. No. Additive Quantity Price Quantity    

B99110

 
bd
B299110
Fluid D 500mL Pack of 10 Each for $461.38
Description & Specifications

Specifications

Certifications/Compliance ISO/DIS, 11014
Format Liquid
For Use With (Application) Sterility testing, non-sterile product testing, food, dairy and Water testing
Packaging Bottle
Shelf Life 365 Days
Color Nearly colorless, clear solution

  • Used for diluting or rinsing samples containing lecithin or oil when performing sterility testing
  • Fluid D contains polysorbate 80, which acts as a surfactant to break down the lecithin or oils present
  • Contains peptic digest of animal tissue, which provides source of nitrogen for bacteria
  • Provided as prepared, ready-to-use diluents in a variety of bottle sizes and closures