Please login to your online account to display your discounted pricing

BD Difco™ Peptone Water


For cultivation of nonfastidious organisms and studying carbohydrate fermentation

Manufacturer: bd  B299110

 View more versions of this product

Catalog No. B99110

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Color Nearly colorless, clear solution
Quantity 500mL
Storage Requirements 15-30°C
Certifications/Compliance ISO/DIS, 11014
Format Liquid
Packaging Bottle
For Use With (Application) Sterility testing, non-sterile product testing, food, dairy and Water testing
Shelf Life 365 Days
Additive Fluid D

  • Used for diluting or rinsing samples containing lecithin or oil when performing sterility testing
  • Fluid D contains polysorbate 80, which acts as a surfactant to break down the lecithin or oils present
  • Contains peptic digest of animal tissue, which provides source of nitrogen for bacteria
  • Provided as prepared, ready-to-use diluents in a variety of bottle sizes and closures

Accessories

365 Days