Please login to your online account to display your discounted pricing

BD Media Supplement Fluid A


For diluting and rinsing in sterility testing

Manufacturer: bd  B299111

 View more versions of this product

Catalog No. B99111

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Storage Requirements 15-30°C
Packaging Bottle
Certifications/Compliance ISO/DIS, 11014
Quantity 500mL
Shelf Life 365 Days
Format Liquid
Additive Fluid A
For Use With (Application) Sterility testing, non-sterile product testing, food, dairy and Water testing
Accessories

365 Days