Please login to your online account to display your discounted pricing
BD Macro-Vue™ RPR (Rapid Plasma Reagin) 18mm Circle Card Tests: Cards
Nontreponemal testing procedure for the serologic detection of syphilis
Manufacturer: bd  B275005
 View more versions of this product
 Alert
RPR Card Antigen Suspension contains sodium azide, which is harmful if swallowed. Contact with acids liberates very toxic gas. After contact with skin, wash immediately with plenty of water. Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. On disposal, flush with a large volume of water to prevent azide buildup.
Catalog No. B875005
  • / Each
Description & Specifications
Specifications
Product Type RPR Card Test Kit
For Use With (Application) Serologic detection of Syphilis
Formulation Cardiolipin 0.003%, Lecithin 0.020 to 0.022%, Cholesterol 0.09%, EDTA 0.0125M, Na2HPO4 0.01M, KH2PO4 0.01M, Sodium Azide 0.2%, Charcoal 0.02%, Choline Chloride 10% w/v, Deionized/Distilled Water
Quantity 500 Tests
Detectable Analytes Detects reagin present in Serum of syphilitic patients
Includes Ampules Antigen 3 x 3mL, 20g dispensing needle, Antigen dispensing bottle, 50 cards with ten 18mm circle spots each