BD Luer-Lok™ PrecisionGlide™ Disposable Syringes with Detachable Needles: 5cc
Sterile, disposable syringes
Manufacturer:  BD 309635
 View more versions of this product
Catalog No. 14-827-49
Description
Description
  • For easy identification of needle gauge, needle package and hub are color coded: 25G, blue; 24G, white; 23G, turquoise; 22G, black; 21G, green; 20G, yellow
  • All syringes are sealed in an Easy Peel™ package
  • Autoclavable out of package
  • With detachable PrecisionGlide™ needle
Syringes with two capacities listed are dual scale.
Specifications
Specifications
Healthcare
Yellow
Yes
1.5 in.
38 mm
Stainless Steel
Syringe
Removable
20 G
1/5 cc
1.5 in.
38 mm
Sterile
5 mL
Documents
Accessories