BD Luer-Lok™ PrecisionGlide™ Disposable Syringes with Detachable Needles: 3cc
Sterile, disposable syringes
Manufacturer:  BD B309571
 View more versions of this product
Catalog No. 14-823-37
Description
Description
Sterile, disposable syringes
  • For easy identification of needle gauge, needle package and hub are color coded: 25G, blue; 24G, white; 23G, turquoise; 22G, black; 21G, green; 20G, yellow
  • All syringes are sealed in an Easy Peel™ package
  • Autoclavable out of package
  • With detachable PrecisionGlide™ needle
  • Stainless-steel needle with polypropylene hub and Luer-Lok™ tip Cat. Nos. 14-826-11 and 14-826-7 are intramuscular needles with a thin-wall design
Specifications
Specifications
Removable
23 G
1/10cc
1 in.
1 in.
25mm
Syringe
Turquoise
3mL
Yes
25mm
Stainless steel
Sterile
Documents
Accessories