Please login to your online account to display your discounted pricing

BD Liquid Amies Elution Swab (Eswab) Collection/Transport System


Intended for the collection and transport of clinical specimens containing aerobes, anaerobes and fastidious bacteria from the collection site to the testing laboratory

Manufacturer: bd  B220532

 View more versions of this product

Catalog No. B220532

  • / Pack of 50 Each
 

Description & Specifications

Specifications

Product Type Elution Swab Collection Kit
For Use With (Application) For the collection and transport of clinical specimens containing aerobes, anaerobes and fastidious Bacteria
Includes Includes Polypropylene screw cap tube with 1mL of liquid Amies medium with a Blue cap, one flexible minitip swab with flocked Nylon Fiber tip
Sterile Yes
Quantity 50 Pack
Swab Configuration Flexible minitip flocked
Storage Requirements 5 to 25°C