Please login to your online account to display your discounted pricing
BD Intramedia Polyethylene Tubing
$24.75 - $238.55
Specifications
Material Polyethylene
Sterile No
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Inner Diameter (English) Outer Diameter (Metric) Inner Diameter (Metric) Outer Price Quantity    
1417011C
  Accessories
bd
427415
0.03 in. 0.048 in. 0.76mm 1.22mm Each for $24.75
1417011H
bd
427450
0.066 in. 0.095 in. 1.67mm 2.42mm Each for $29.85
1417012J
  Accessories
bd
427441
0.055 in. 0.075 in. 1.4mm 1.9mm Each for $238.55
1417012N
  Accessories
bd
427426
0.034 in. 0.06 in. 0.86mm 1.52mm Each for $186.80
Description & Specifications
Specifications
Material Polyethylene
Sterile No