missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BD Integra™ Syringe

Designed to have low waste space to help reduce medication waste

Manufacturer:  BD 305274

 View more versions of this product

Catalog No. 1482355



Description

Description

  • 3mL
  • Sterile
  • Retractable needle
  • Shelf Life: 5 years

Specifications

Specifications

Retractible and detachable
3mL
38mm
Syringe
21 G
1.5 in.
Sterile
Documents