missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BD Integra™ Syringe
Designed to have low waste space to help reduce medication waste
Supplier: BD 305271
Description
- 3mL
- Sterile
- Retractable needle
- Shelf Life: 5 years
Specifications
23 G | |
Syringe | |
Retractible and Detachable |
1 in. | |
3 mL | |
Sterile |