Please login to your online account to display your discounted pricing

BD General Use Syringes


For general use

Manufacturer: bd  309647

 View more versions of this product

Catalog No. B309647

  • / Pack of 125 Each
  • / Case of 4 Pack
 

Description & Specifications

Specifications

Disposable Yes
Volume (Metric) 5mL
Connection Type Slip-Tip
Graduations 0.5mL
Termination Slip tip
Capacity (Metric) 5mL
Sterile Yes
Type Syringe